Product Details

SNP ID
rs6499322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70581439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTATTTGAAGTTATTGTTTTTT[C/T]CCCCCTTTTCCTTGGAGTGTATTCT
Phenotype
MIM: 612081 MIM: 605592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL34 PubMed Links

Gene Details

Gene
IL34
Gene Name
interleukin 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172771.1 Intron NP_001166242.1
NM_001172772.1 Intron NP_001166243.1
NM_152456.2 Intron NP_689669.2
XM_011522902.2 Intron XP_011521204.1
XM_011522903.2 Intron XP_011521205.1
Gene
SF3B3
Gene Name
splicing factor 3b subunit 3
There are no transcripts associated with this gene.

View Full Product Details