Product Details

SNP ID
rs9892555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37518816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGATGACGGGGGCCCCGTCCCT[C/T]GCGGTGTTCTTCATATCGATTCAGG
Phenotype
MIM: 606618 MIM: 607291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP14 PubMed Links

Gene Details

Gene
DUSP14
Gene Name
dual specificity phosphatase 14
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163544.2 4276 UTR 3 NP_001157016.1
NM_001163545.2 4276 UTR 3 NP_001157017.1
NM_001163546.2 4276 UTR 3 NP_001157018.1
NM_001163547.2 4276 UTR 3 NP_001157019.1
NM_007247.5 4276 UTR 3 NP_009178.3
NM_080550.4 4276 UTR 3 NP_542117.3
NM_198882.2 4276 UTR 3 NP_942583.1
XM_005256980.4 4276 UTR 3 XP_005257037.1
XM_017024076.1 4276 UTR 3 XP_016879565.1
XM_017024077.1 4276 UTR 3 XP_016879566.1
XM_017024078.1 4276 UTR 3 XP_016879567.1
XM_017024079.1 4276 UTR 3 XP_016879568.1
XM_017024080.1 4276 UTR 3 XP_016879569.1
XM_017024081.1 4276 UTR 3 XP_016879570.1
XM_017024082.1 4276 UTR 3 XP_016879571.1
XM_017024083.1 4276 UTR 3 XP_016879572.1
XM_017024084.1 4276 UTR 3 XP_016879573.1
XM_017024085.1 4276 UTR 3 XP_016879574.1
XM_017024086.1 4276 UTR 3 XP_016879575.1
XM_017024087.1 4276 UTR 3 XP_016879576.1
XM_017024088.1 4276 UTR 3 XP_016879577.1
XM_017024089.1 4276 UTR 3 XP_016879578.1
XM_017024090.1 4276 UTR 3 XP_016879579.1
XM_017024091.1 4276 UTR 3 XP_016879580.1
XM_017024092.1 4276 UTR 3 XP_016879581.1
XM_017024093.1 4276 UTR 3 XP_016879582.1
XM_017024094.1 4276 UTR 3 XP_016879583.1
XM_017024095.1 4276 UTR 3 XP_016879584.1
XM_017024096.1 4276 Intron XP_016879585.1
XM_017024097.1 4276 UTR 3 XP_016879586.1
XM_017024098.1 4276 UTR 3 XP_016879587.1
XM_017024099.1 4276 UTR 3 XP_016879588.1
XM_017024100.1 4276 Intron XP_016879589.1
XM_017024101.1 4276 Intron XP_016879590.1
XM_017024102.1 4276 UTR 3 XP_016879591.1
XM_017024103.1 4276 UTR 3 XP_016879592.1
XM_017024104.1 4276 UTR 3 XP_016879593.1
XM_017024105.1 4276 Intron XP_016879594.1
XM_017024106.1 4276 UTR 3 XP_016879595.1

View Full Product Details