Product Details
- SNP ID
-
rs6127140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:54214378 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAGAAGATGTCAATGTTACTTTC[C/G]AAGATCAACAAAAGATAAACAAATT
- Phenotype
-
MIM: 604898
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PFDN4
PubMed Links
Gene Details
- Gene
- PFDN4
- Gene Name
- prefoldin subunit 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002623.3 |
190 |
Missense Mutation |
CAA,GAA |
Q18E |
NP_002614.2 |
View Full Product Details