Product Details
- SNP ID
-
rs9298197
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:72022156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGAATATGTGATGGAGCCATGTT[G/T]TTTTCAAATCTCAAACAAAGGCTCA
- Phenotype
-
MIM: 604775
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MSC-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142866053,rs78580746] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MSC-AS1
- Gene Name
- MSC antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- TRPA1
- Gene Name
- transient receptor potential cation channel subfamily A member 1
View Full Product Details