Product Details

SNP ID
rs4148310
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.4:69589432 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
ATTTTTGACAGGAAAACAAACAACA[C/T]TGTATGACCAAAAATATAGCCGTTG
Phenotype
MIM: 604716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT2A1 PubMed Links

Gene Details

Gene
UGT2A1
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252274.2 1421 Silent Mutation CAA,CAG Q674Q NP_001239203.2
NM_001252275.2 1421 Silent Mutation CAA,CAG Q508Q NP_001239204.2
NM_001301239.1 1421 Silent Mutation CAA,CAG Q464Q NP_001288168.1
NM_006798.4 1421 Silent Mutation CAA,CAG Q508Q NP_006789.3
Gene
UGT2A2
Gene Name
UDP glucuronosyltransferase family 2 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105677.2 1421 Silent Mutation CAA,CAG Q517Q NP_001099147.2
NM_001301233.1 1421 Silent Mutation CAA,CAG Q473Q NP_001288162.1

View Full Product Details