Product Details

SNP ID
rs3811127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136207754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTGAAGAGCAGACGACAGGGGG[C/G]GCTTTAGAAGTCTCCCTGGGGCCTA
Phenotype
MIM: 600577 MIM: 612860
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LHX3 PubMed Links

Gene Details

Gene
LHX3
Gene Name
LIM homeobox 3
There are no transcripts associated with this gene.

Gene
QSOX2
Gene Name
quiescin sulfhydryl oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181701.3 3109 UTR 3 NP_859052.3

View Full Product Details