Product Details

SNP ID
rs12789846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1739358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGGGTTGTTGGTGTAGTGTA[A/G]ACCACTGGATGGTAGATGGGTAGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFITM10 PubMed Links

Gene Details

Gene
IFITM10
Gene Name
interferon induced transmembrane protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170820.3 Intron NP_001164291.2

View Full Product Details