Product Details

SNP ID
rs11100679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140864005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTATGGTAGCCTCAGGGATGTCA[C/G]GGCAGTAACTGAAATGGCCACTGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF150 PubMed Links

Gene Details

Gene
RNF150
Gene Name
ring finger protein 150
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020724.1 5111 Intron NP_065775.1
XM_005263150.4 5111 UTR 3 XP_005263207.1
XM_011532147.2 5111 UTR 3 XP_011530449.1
XM_011532148.2 5111 UTR 3 XP_011530450.1
XM_017008475.1 5111 UTR 3 XP_016863964.1
XM_017008476.1 5111 UTR 3 XP_016863965.1

View Full Product Details