Product Details

SNP ID
rs4777502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72345454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCACAGCTTGCTTACCTCAGCAA[C/T]TCACAGCGGAAGTGTGACAAACGTT
Phenotype
MIM: 606869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HEXA PubMed Links

Gene Details

Gene
HEXA
Gene Name
hexosaminidase subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000520.5 2052 Silent Mutation GAC,GAT D506D NP_000511.2
NM_001318825.1 2052 Silent Mutation GAC,GAT D517D NP_001305754.1

View Full Product Details