Product Details

SNP ID
rs12185514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57874013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACACTTGAAAGGTCTTTCTCC[A/G]GCGTGAACTCGTTGATGGCTCCTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2CP5 PubMed Links

Gene Details

Gene
UBE2CP5
Gene Name
ubiquitin conjugating enzyme E2 C pseudogene 5
There are no transcripts associated with this gene.

Gene
ZNF587
Gene Name
zinc finger protein 587
There are no transcripts associated with this gene.

Gene
ZNF814
Gene Name
zinc finger protein 814
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144989.1 Intron NP_001138461.1
XM_017027197.1 Intron XP_016882686.1

View Full Product Details