Product Details

SNP ID
rs7831847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:140513854 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAAAATGCCTGCTTCTCCGCCA[A/G]TTTTGATACTGGCTTTGCTTTCTTT
Phenotype
MIM: 606229 MIM: 607268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGO2 PubMed Links

Gene Details

Gene
AGO2
Gene Name
argonaute 2, RISC catalytic component
There are no transcripts associated with this gene.

Gene
CHRAC1
Gene Name
chromatin accessibility complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017444.5 Intron NP_059140.1

View Full Product Details