Product Details
- SNP ID
-
rs11166118
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:92080560 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTACTGGGCCTCCAGGGCGTCGTA[C/T]CTCTGTGAGACATGGCTCGCTGTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BTBD8
PubMed Links
Gene Details
- Gene
- BTBD8
- Gene Name
- BTB domain containing 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183242.3 |
256 |
UTR 5 |
|
|
NP_899065.2 |
- Gene
- SETSIP
- Gene Name
- SET-like protein
There are no transcripts associated with this gene.
View Full Product Details