Product Details

SNP ID
rs11668687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43979188 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATTGTATAGAATTTTACAAAACT[A/G]GGAACACACTAAGCATACTTGCTCC
Phenotype
MIM: 604086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF155 PubMed Links

Gene Details

Gene
ZNF155
Gene Name
zinc finger protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260486.1 Intron NP_001247415.1
NM_001260487.1 Intron NP_001247416.1
NM_001260488.1 Intron NP_001247417.1
NM_003445.3 Intron NP_003436.3
NM_198089.2 Intron NP_932355.2
XM_005259215.3 Intron XP_005259272.1
XM_011527278.2 Intron XP_011525580.1
XM_011527279.2 Intron XP_011525581.1
XM_017027248.1 Intron XP_016882737.1
Gene
ZNF221
Gene Name
zinc finger protein 221
There are no transcripts associated with this gene.

View Full Product Details