Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300776.1 | Intron | NP_001287705.1 | |||
NM_001300777.1 | Intron | NP_001287706.1 | |||
NM_001300778.1 | Intron | NP_001287707.1 | |||
NM_003440.3 | Intron | NP_003431.2 | |||
XM_005266188.4 | Intron | XP_005266245.1 | |||
XM_011534833.2 | Intron | XP_011533135.1 | |||
XM_011534834.2 | Intron | XP_011533136.1 | |||
XM_011534835.2 | Intron | XP_011533137.1 | |||
XM_011534836.2 | Intron | XP_011533138.1 | |||
XM_011534837.2 | Intron | XP_011533139.1 | |||
XM_011534838.2 | Intron | XP_011533140.1 | |||
XM_011534839.2 | Intron | XP_011533141.1 | |||
XM_011534840.2 | Intron | XP_011533142.1 | |||
XM_017019924.1 | Intron | XP_016875413.1 | |||
XM_017019925.1 | Intron | XP_016875414.1 |