Product Details

SNP ID
rs12299933
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133056693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAACCTTTTTTCTTTGCTTTTC[A/T]TGTGTTTTTCATATTCCCCATTTAA
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF84 PubMed Links

Gene Details

Gene
ZNF84
Gene Name
zinc finger protein 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127372.2 Intron NP_001120844.1
NM_001289971.1 Intron NP_001276900.1
NM_001289972.1 Intron NP_001276901.1
NM_003428.5 Intron NP_003419.3
XM_005266185.1 Intron XP_005266242.1
XM_005266186.1 Intron XP_005266243.1
XM_011534832.1 Intron XP_011533134.1

View Full Product Details