Product Details

SNP ID
rs11003860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:53803253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATTAGCACTTTTTGCCTATTTT[A/G]CTAATTAAATTCAAGTGGTATATAA
Phenotype
MIM: 605514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDH15 PubMed Links

Gene Details

Gene
PCDH15
Gene Name
protocadherin related 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142763.1 8770 Intron NP_001136235.1
NM_001142764.1 8770 Intron NP_001136236.1
NM_001142765.1 8770 Intron NP_001136237.1
NM_001142766.1 8770 Intron NP_001136238.1
NM_001142767.1 8770 Intron NP_001136239.1
NM_001142768.1 8770 Intron NP_001136240.1
NM_001142769.1 8770 Intron NP_001136241.1
NM_001142770.1 8770 Intron NP_001136242.1
NM_001142771.1 8770 UTR 3 NP_001136243.1
NM_001142772.1 8770 UTR 3 NP_001136244.1
NM_001142773.1 8770 Intron NP_001136245.1
NM_033056.3 8770 Intron NP_149045.3
XM_017016571.1 8770 Intron XP_016872060.1
XM_017016572.1 8770 UTR 3 XP_016872061.1
XM_017016573.1 8770 Intron XP_016872062.1

View Full Product Details