Product Details
- SNP ID
-
rs7563454
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231201259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGCTTCCAGATCACTACCCACTC[C/T]TTCACAAACCCCAGCTCCTTTGAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARMC9
PubMed Links
Gene Details
- Gene
- ARMC9
- Gene Name
- armadillo repeat containing 9
View Full Product Details