Product Details

SNP ID
rs10799659
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:20664412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCGAGGGGCTCCTGCCCAGGGC[A/G]GAGGTGGGCTCTCTGGGGAACAGGG
Phenotype
MIM: 602202 MIM: 605037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDOST PubMed Links

Gene Details

Gene
DDOST
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
There are no transcripts associated with this gene.

Gene
KIF17
Gene Name
kinesin family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122819.2 3570 UTR 3 NP_001116291.1
NM_001287212.1 3570 UTR 3 NP_001274141.1
NM_020816.3 3570 UTR 3 NP_065867.2
XM_005245950.3 3570 Intron XP_005246007.1
XM_005245951.3 3570 Intron XP_005246008.1
XM_011541837.1 3570 Intron XP_011540139.1
XM_011541838.1 3570 Intron XP_011540140.1
XM_011541839.2 3570 Intron XP_011540141.1
XM_011541840.2 3570 Intron XP_011540142.1
XM_011541841.2 3570 Intron XP_011540143.1
XM_011541842.1 3570 Intron XP_011540144.1
XM_011541843.1 3570 Intron XP_011540145.1
XM_011541844.2 3570 Intron XP_011540146.1
XM_011541845.2 3570 Intron XP_011540147.1
XM_011541846.2 3570 Intron XP_011540148.1
XM_011541847.2 3570 Intron XP_011540149.1

View Full Product Details