Product Details

SNP ID
rs12823489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21021270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAATTATCTGTCTTAGTAGATG[C/G]TCCCATTCCCGCCCCACCCCCACCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLCO1B7 PubMed Links

Gene Details

Gene
SLCO1B7
Gene Name
solute carrier organic anion transporter family member 1B7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009562.4 Intron NP_001009562.3

View Full Product Details