Product Details
- SNP ID
-
hCV31200741
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:116535314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGCTTTGAGAAAATAAAATGGCC[A/C]CCTTGCTCCGCTGTTCTTTCTTTGT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00173
PubMed Links
Gene Details
- Gene
- LINC00173
- Gene Name
- long intergenic non-protein coding RNA 173
There are no transcripts associated with this gene.
View Full Product Details