Product Details

SNP ID
rs10850661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116535690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTGGTGTCAAGTGGGAGCTCA[A/G]AAACTGAGGGGCAGGCAGATGCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00173 PubMed Links

Gene Details

Gene
LINC00173
Gene Name
long intergenic non-protein coding RNA 173
There are no transcripts associated with this gene.

View Full Product Details