Product Details

SNP ID
rs11247857
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:25618250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACTTCAACGCATTCCGGAGCCG[C/T]CTCCGCCACCCGGTCCTGGGAACGA
Phenotype
MIM: 616772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAN1C1 PubMed Links

Gene Details

Gene
MAN1C1
Gene Name
mannosidase alpha class 1C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289010.1 783 Silent Mutation CGC,CGT R151R NP_001275939.1
NM_020379.3 783 Silent Mutation CGC,CGT R151R NP_065112.1
XM_017001860.1 783 UTR 5 XP_016857349.1
XM_017001861.1 783 Silent Mutation CGC,CGT R151R XP_016857350.1

View Full Product Details