Product Details

SNP ID
rs12190454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37258920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCTACCTTTTCTAATGGATTCA[A/G]GTTTTTTCCTTCTGGGATAATCTTT
Phenotype
MIM: 616880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D22B PubMed Links

Gene Details

Gene
TBC1D22B
Gene Name
TBC1 domain family member 22B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017772.3 115 Intron NP_060242.2
XM_011514738.2 115 Intron XP_011513040.1
XM_011514739.1 115 Intron XP_011513041.1
Gene
TMEM217
Gene Name
transmembrane protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162900.1 115 Intron NP_001156372.1
NM_001286401.1 115 Intron NP_001273330.1
NM_145316.3 115 Intron NP_660359.2
XM_011514362.1 115 Intron XP_011512664.1
XM_011514363.2 115 Intron XP_011512665.1
XM_011514364.2 115 Intron XP_011512666.1
XM_011514365.2 115 UTR 5 XP_011512667.1
XM_011514366.2 115 Intron XP_011512668.1
XM_011514367.2 115 Intron XP_011512669.1
XM_011514369.2 115 Intron XP_011512671.1
XM_011514370.2 115 Intron XP_011512672.1

View Full Product Details