Product Details

SNP ID
rs11141015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:85544213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATAAAAAGCTTTATTCAGGCTCA[A/T]AATGTATGCTAAAGTACTTCATTAA
Phenotype
MIM: 606830
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AGTPBP1 PubMed Links

Gene Details

Gene
AGTPBP1
Gene Name
ATP/GTP binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286715.1 6811 Intron NP_001273644.1
NM_001286717.1 6811 Intron NP_001273646.1
NM_015239.2 6811 Intron NP_056054.2
XM_005251847.3 6811 UTR 3 XP_005251904.1
XM_005251848.2 6811 Intron XP_005251905.1
XM_005251849.3 6811 Intron XP_005251906.1
XM_011518418.2 6811 Intron XP_011516720.1
XM_011518420.2 6811 Intron XP_011516722.1
XM_011518421.2 6811 Intron XP_011516723.1
XM_017014544.1 6811 Intron XP_016870033.1
XM_017014545.1 6811 Intron XP_016870034.1
XM_017014546.1 6811 Intron XP_016870035.1
XM_017014547.1 6811 Intron XP_016870036.1
XM_017014548.1 6811 Intron XP_016870037.1
XM_017014549.1 6811 Intron XP_016870038.1
XM_017014550.1 6811 Intron XP_016870039.1
Gene
LOC101927603
Gene Name
uncharacterized LOC101927603
There are no transcripts associated with this gene.

View Full Product Details