Product Details

SNP ID
rs12820185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27780480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCAGGAGGCCGGCGGCCCGGA[A/G]GTGCAGCAGCTGCGCGGCCTCAGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL42 PubMed Links

Gene Details

Gene
KLHL42
Gene Name
kelch like family member 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020782.1 524 Silent Mutation GAA,GAG E50E NP_065833.1
XM_017019698.1 524 Silent Mutation GAA,GAG E50E XP_016875187.1
Gene
MANSC4
Gene Name
MANSC domain containing 4
There are no transcripts associated with this gene.

View Full Product Details