Product Details

SNP ID
rs12602765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1831274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATCCCTATCTCCACGGATGGC[A/C]CCATTATCTATTCAGTTCAGTGCTA
Phenotype
MIM: 179835
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPA1 PubMed Links

Gene Details

Gene
RPA1
Gene Name
replication protein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002945.3 Intron NP_002936.1
Gene
SMYD4
Gene Name
SET and MYND domain containing 4
There are no transcripts associated with this gene.

View Full Product Details