Product Details

SNP ID
rs11570140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11965475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAGTAAGTAATATTGCTTGAAC[A/G]ATTATTCATTGGTGTGAACTATTCT
Phenotype
MIM: 109545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF17 PubMed Links

Gene Details

Gene
TNFRSF17
Gene Name
TNF receptor superfamily member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001192.2 Intron NP_001183.2

View Full Product Details