Product Details

SNP ID
rs11264565
Assay Type
Functionally Tested
NCBI dbSNP Submissions
41
Location
Chr.1:156827767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTGAAATAGAAGTGAGAGACT[A/C]TGAACCTGAAATGGTCAGCTTCTGG
Phenotype
MIM: 191315
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NTRK1 PubMed Links

Gene Details

Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007792.1 Intron NP_001007793.1
NM_001012331.1 Intron NP_001012331.1
NM_002529.3 Intron NP_002520.2

View Full Product Details