Product Details

SNP ID
rs11581406
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:115654189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTAGGAGGCGGGTTGGGGTTG[G/T]AGGGGGACCATGCTCTTTGGTAATT
Phenotype
MIM: 610132
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VANGL1 PubMed Links

Gene Details

Gene
VANGL1
Gene Name
VANGL planar cell polarity protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172411.1 Intron NP_001165882.1
NM_001172412.1 Intron NP_001165883.1
NM_138959.2 Intron NP_620409.1

View Full Product Details