Product Details

SNP ID
rs12891251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:56582777 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCGGGAAGCTCGAGGTTTTTTTA[C/G]TATTCCCTTCCTCTTTATCACAATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM260 PubMed Links

Gene Details

Gene
TMEM260
Gene Name
transmembrane protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017799.3 Intron NP_060269.3
XM_005267771.1 Intron XP_005267828.1
XM_006720176.1 Intron XP_006720239.1
XM_006720178.1 Intron XP_006720241.1
XM_011536851.1 Intron XP_011535153.1
XM_011536852.1 Intron XP_011535154.1
XM_011536853.1 Intron XP_011535155.1
XM_017021379.1 Intron XP_016876868.1
XM_017021380.1 Intron XP_016876869.1

View Full Product Details