Product Details

SNP ID
rs12936722
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67340370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACTCTATCCCAAATGTTCTTG[G/T]ATGCTCTATGTTGCTTTTCTATTCT
Phenotype
MIM: 604450
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
PSMD12 PubMed Links

Gene Details

Gene
PSMD12
Gene Name
proteasome 26S subunit, non-ATPase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316341.1 2236 UTR 3 NP_001303270.1
NM_002816.4 2236 UTR 3 NP_002807.1
NM_174871.3 2236 UTR 3 NP_777360.1

View Full Product Details