Product Details

SNP ID
rs11583363
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:196979317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATACATCACTTAATGATAGGAAA[A/C]CGTTCTGAGAAGTGTGTCTATAGGC
Phenotype
MIM: 608593
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFHR5 PubMed Links

Gene Details

Gene
CFHR5
Gene Name
complement factor H related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030787.3 Intron NP_110414.1
XM_011510020.2 Intron XP_011508322.1

View Full Product Details