Product Details

SNP ID
rs11965276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57050905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTTTCTTATTAAGTGCAATAG[A/T]GTGTTGGTGGTAGCTTGAACTGGAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1586 PubMed Links

Gene Details

Gene
KIAA1586
Gene Name
KIAA1586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286274.1 Intron NP_001273203.1
NM_001286275.1 Intron NP_001273204.1
NM_001286276.1 Intron NP_001273205.1
NM_020931.3 Intron NP_065982.1
XM_011514767.2 Intron XP_011513069.1

View Full Product Details