Product Details

SNP ID
rs7280551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36037311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATATACAATTTTTACTGACCAAT[A/T]AAAAATAAAGAGGCCAGGTGCAGTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SETD4 PubMed Links

Gene Details

Gene
SETD4
Gene Name
SET domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007259.2 Intron NP_001007260.1
NM_001007261.2 Intron NP_001007262.1
NM_001286752.1 Intron NP_001273681.1
NM_017438.4 Intron NP_059134.1
XM_011529636.1 Intron XP_011527938.1
XM_011529637.1 Intron XP_011527939.1
XM_011529638.2 Intron XP_011527940.1
XM_011529639.1 Intron XP_011527941.1
XM_011529640.2 Intron XP_011527942.1
XM_011529642.1 Intron XP_011527944.1
XM_011529643.1 Intron XP_011527945.1
XM_011529644.1 Intron XP_011527946.1
XM_017028403.1 Intron XP_016883892.1
XM_017028404.1 Intron XP_016883893.1
XM_017028405.1 Intron XP_016883894.1

View Full Product Details