Product Details

SNP ID
rs11037737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43924233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAGTACCCAGAGGATTTATCA[G/T]ATCTGGGATCTGGTGTCCGCAGTGG
Phenotype
MIM: 610603
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACCSL PubMed Links

Gene Details

Gene
ACCSL
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031854.2 Intron NP_001027025.2
XM_017017714.1 Intron XP_016873203.1
XM_017017715.1 Intron XP_016873204.1
Gene
ALKBH3
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
There are no transcripts associated with this gene.

Gene
ALKBH3-AS1
Gene Name
ALKBH3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details