Product Details

SNP ID
rs12911708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64748729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGGCAAGGAAGCCACGTTAAAT[C/T]GGCCTGTCATGGTGTGCATCCTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBPMS2 PubMed Links

Gene Details

Gene
RBPMS2
Gene Name
RNA binding protein with multiple splicing 2
There are no transcripts associated with this gene.

View Full Product Details