Product Details

SNP ID
rs11011046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:37130162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAATTTATAATAATTTATATTA[C/T]AAATTATACAGTTTACAATAATTCT
Phenotype
MIM: 610856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD30A PubMed Links

Gene Details

Gene
ANKRD30A
Gene Name
ankyrin repeat domain 30A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052997.2 Intron NP_443723.2
XM_011519757.2 Intron XP_011518059.1
XM_011519758.2 Intron XP_011518060.1
XM_017016883.1 Intron XP_016872372.1

View Full Product Details