Product Details

SNP ID
rs12472986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72912848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGATGTTTGTTTCTGGGTAGTA[G/T]GATTTGACAATTATAAGATCTCTGT
Phenotype
MIM: 600034
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EMX1 PubMed Links

Gene Details

Gene
EMX1
Gene Name
empty spiracles homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004097.2 1702 Intron NP_004088.2
XM_005264203.3 1702 Intron XP_005264260.1
XM_011532697.2 1702 UTR 5 XP_011530999.1

View Full Product Details