Product Details

SNP ID
rs13015457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69898631 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAATGGAATGGGGTAATCTCAGT[G/T]GCTGTAAGTTTCTTTCTAACTAATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNRNP27 PubMed Links

Gene Details

Gene
SNRNP27
Gene Name
small nuclear ribonucleoprotein U4/U6.U5 subunit 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006857.2 Intron NP_006848.1

View Full Product Details