Product Details
- SNP ID
-
rs13011500
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189441494 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCATTCCGCTACTGCGCAAAGA[A/T]GGTGGAGGAGGAGAACATCCGCGTG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR75
PubMed Links
Gene Details
- Gene
- WDR75
- Gene Name
- WD repeat domain 75
View Full Product Details