Product Details

SNP ID
rs10931429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189665692 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGTTATTAACATGTGCTTTAGA[A/G]TTTTAAACAGGTCTAAGTATGACAA
Phenotype
MIM: 609803
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKAR PubMed Links

Gene Details

Gene
ANKAR
Gene Name
ankyrin and armadillo repeat containing
There are no transcripts associated with this gene.

Gene
ASNSD1
Gene Name
asparagine synthetase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019048.2 510 Intron NP_061921.1
XM_017004381.1 510 Intron XP_016859870.1
XM_017004382.1 510 UTR 5 XP_016859871.1

View Full Product Details