Product Details
- SNP ID
-
rs12845571
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:49118294 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTGACCTACAGAAACAGCAATT[T/G]CATTTGGTTCAACCTAATAACATGC
- Phenotype
-
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPKOW
PubMed Links
Gene Details
- Gene
- GPKOW
- Gene Name
- G-patch domain and KOW motifs
View Full Product Details