Product Details

SNP ID
rs12106596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38660972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAACCTCTTTCTCTTCTCCAGCA[A/G]AAACAGTGCCAGGGCCTTACAATGT
Phenotype
MIM: 607757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBY1 PubMed Links

Gene Details

Gene
CBY1
Gene Name
chibby family member 1, beta catenin antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002880.1 Intron NP_001002880.2
NM_015373.3 Intron NP_056188.1
Gene
FAM227A
Gene Name
family with sequence similarity 227 member A
There are no transcripts associated with this gene.

View Full Product Details