Product Details

SNP ID
rs7020130
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93100259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTGAGTCTGTGCCTCAAGGAGT[A/G]AACCCAAGAGGGGGCCCCACTGTGT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CARD19 PubMed Links

Gene Details

Gene
CARD19
Gene Name
caspase recruitment domain family member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318010.1 Intron NP_001304939.1
NM_001318011.1 Intron NP_001304940.1
NM_032310.4 Intron NP_115686.3

View Full Product Details