Product Details

SNP ID
rs11666067
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1389518 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTCGGACCCTGCTGGAGATGAC[A/C]CCTCCGTTTCCGTCATTCTCTTTCT
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
There are no transcripts associated with this gene.

Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024407.4 1164 Intron NP_077718.3
XM_005259556.4 1164 Intron XP_005259613.2
XM_017026768.1 1164 Missense Mutation ACC,CCC T355P XP_016882257.1

View Full Product Details