Product Details

SNP ID
rs10915744
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:224929112 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCAATAAATATTGTAGTTGTTGC[A/G]TGAGTGAAATATAGAACATTAACTT
Phenotype
MIM: 603341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145154.1 690 Intron NP_001138626.1
NM_001373.1 690 Intron NP_001364.1
NM_144989.2 690 Intron NP_659426.2
XM_006711735.3 690 UTR 5 XP_006711798.1
XM_011544057.2 690 UTR 5 XP_011542359.1
XM_011544058.2 690 UTR 5 XP_011542360.1
XM_011544059.2 690 UTR 5 XP_011542361.1
XM_011544061.2 690 UTR 5 XP_011542363.1
XM_011544062.2 690 UTR 5 XP_011542364.1
XM_011544063.2 690 UTR 5 XP_011542365.1
XM_011544064.2 690 UTR 5 XP_011542366.1
XM_011544065.2 690 UTR 5 XP_011542367.1
XM_011544066.2 690 UTR 5 XP_011542368.1
XM_011544067.2 690 UTR 5 XP_011542369.1
XM_011544069.2 690 UTR 5 XP_011542371.1
XM_011544070.2 690 UTR 5 XP_011542372.1
XM_011544071.2 690 UTR 5 XP_011542373.1
XM_011544072.2 690 UTR 5 XP_011542374.1
XM_011544073.2 690 Intron XP_011542375.1
XM_011544074.2 690 Intron XP_011542376.1
XM_011544076.1 690 Intron XP_011542378.1
XM_011544079.1 690 Intron XP_011542381.1
XM_011544081.1 690 Intron XP_011542383.1
XM_017000295.1 690 UTR 5 XP_016855784.1
XM_017000296.1 690 UTR 5 XP_016855785.1
XM_017000297.1 690 UTR 5 XP_016855786.1
XM_017000298.1 690 UTR 5 XP_016855787.1
XM_017000299.1 690 Intron XP_016855788.1
XM_017000300.1 690 UTR 5 XP_016855789.1

View Full Product Details