Product Details

SNP ID
rs6429447
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:244050793 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCCAACATTTGAGACATGTATA[A/C]CATTATCTCAGCAGAAAAAAATAAA
Phenotype
MIM: 608433
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC339529 PubMed Links

Gene Details

Gene
LOC339529
Gene Name
uncharacterized LOC339529
There are no transcripts associated with this gene.

Gene
ZBTB18
Gene Name
zinc finger and BTB domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278196.1 1464 Intron NP_001265125.1
NM_006352.4 1464 Intron NP_006343.2
NM_205768.2 1464 Intron NP_991331.1
XM_005273006.2 1464 Intron XP_005273063.1
XM_017000060.1 1464 UTR 5 XP_016855549.1

View Full Product Details