Product Details

SNP ID
rs12927376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1979151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGCCCTGCGTGTGACGGTGCA[A/G]CAGCGGCTCTGGATGGCGCCCGGCG
Phenotype
MIM: 600924 MIM: 611256 MIM: 605915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 1399 Silent Mutation TGC,TGT C343C NP_001254650.1
NM_144603.3 1399 Silent Mutation TGC,TGT C338C NP_653204.1
NM_172167.2 1399 Silent Mutation TGC,TGT C339C NP_751907.1
NM_172168.2 1399 Silent Mutation TGC,TGT C344C NP_751908.1
XM_005255099.4 1399 Silent Mutation TGC,TGT C344C XP_005255156.1
XM_017022927.1 1399 Silent Mutation TGC,TGT C344C XP_016878416.1
XM_017022928.1 1399 Silent Mutation TGC,TGT C344C XP_016878417.1
Gene
TBL3
Gene Name
transducin beta like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006453.2 1399 Intron NP_006444.2

View Full Product Details