Product Details

SNP ID
rs11615570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110712320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAACATATAAAATGACAATAT[A/C]CCTTATAGATACACACACATTGAAA
Phenotype
MIM: 611227 MIM: 176914
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HVCN1 PubMed Links

Gene Details

Gene
HVCN1
Gene Name
hydrogen voltage gated channel 1
There are no transcripts associated with this gene.

Gene
PPP1CC
Gene Name
protein phosphatase 1 catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244974.1 Intron NP_001231903.1
NM_002710.3 Intron NP_002701.1
XM_011538504.2 Intron XP_011536806.1
XM_011538505.2 Intron XP_011536807.1

View Full Product Details