Product Details

SNP ID
rs1807130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33573162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAACTAATGTTGTATTTTCTCTG[C/T]GGCAACCCTAGAATGAGTAATTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD18B PubMed Links

Gene Details

Gene
ANKRD18B
Gene Name
ankyrin repeat domain 18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244752.1 Intron NP_001231681.1
XM_011517881.2 Intron XP_011516183.1
XM_011517883.2 Intron XP_011516185.1
XM_011517885.2 Intron XP_011516187.1
XM_017014728.1 Intron XP_016870217.1

View Full Product Details